Order Entry
Italy
ContactUsLinkComponent
2-(Morpholin-4-yl)-2-(pyridin-2-yl)acetonitrile, tech
2-(Morpholin-4-yl)-2-(pyridin-2-yl)acetonitrile, tech
Catalog # APOSOR32972-1G
Supplier:  Apollo Scientific
CAS Number:  
2-(Morpholin-4-yl)-2-(pyridin-2-yl)acetonitrile, tech
Catalog # APOSOR32972-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :203.24 g/mol
Storage Temperature:Ambient
CAS Number:53813-00-6
Formula:C₁₁H₁₃N₃O
MDL Number:MFCD00231856