Order Entry
Italy
ContactUsLinkComponent
(2R,6S)-10-Methoxy-3-methyl-4,8-dioxa-3-azatricyclo[7.4.0.02,6]trideca-1(13),9,11-triene, tech
(2R,6S)-10-Methoxy-3-methyl-4,8-dioxa-3-azatricyclo[7.4.0.02,6]trideca-1(13),9,11-triene, tech
Catalog # APOSOR32971-1G
Supplier:  Apollo Scientific
CAS Number:  
(2R,6S)-10-Methoxy-3-methyl-4,8-dioxa-3-azatricyclo[7.4.0.02,6]trideca-1(13),9,11-triene, tech
Catalog # APOSOR32971-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :221.252 g/mol
Storage Temperature:Ambient
CAS Number:477713-63-6
Formula:C₁₂H₁₅NO₃
MDL Number:MFCD00231686