Order Entry
Italy
ContactUsLinkComponent
2-{[(3-Methoxyphenyl)amino]methylidene}-5,5-dimethylcyclohexane-1,3-dione, tech
2-{[(3-Methoxyphenyl)amino]methylidene}-5,5-dimethylcyclohexane-1,3-dione, tech
Codice catalogo APOSOR32935-1G
Supplier:  Apollo Scientific
CAS Number:  
2-{[(3-Methoxyphenyl)amino]methylidene}-5,5-dimethylcyclohexane-1,3-dione, tech
Codice catalogo APOSOR32935-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :273.327 g/mol
Storage Temperature:Ambient
CAS Number:64869-19-8
Formula:C₁₆H₁₉NO₃
MDL Number:MFCD02936630