Order Entry
Italy
ContactUsLinkComponent
(4,5-Dichloro-6-oxo-1,6-dihydropyridazin-1-yl)methyl acetate, tech
(4,5-Dichloro-6-oxo-1,6-dihydropyridazin-1-yl)methyl acetate, tech
Catalog # APOSOR32920-1G
Supplier:  Apollo Scientific
CAS Number:  
(4,5-Dichloro-6-oxo-1,6-dihydropyridazin-1-yl)methyl acetate, tech
Catalog # APOSOR32920-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :237.04 g/mol
Storage Temperature:Ambient
CAS Number:97137-15-0
Formula:C₇H₆Cl₂N₂O₃
MDL Number:MFCD03305724