Order Entry
Italy
ContactUsLinkComponent
1-[2-(3-Chlorophenyl)-1,3-thiazol-5-yl]ethan-1-one, tech
1-[2-(3-Chlorophenyl)-1,3-thiazol-5-yl]ethan-1-one, tech
Catalog # APOSOR32906-1G
Supplier:  Apollo Scientific
CAS Number:  
1-[2-(3-Chlorophenyl)-1,3-thiazol-5-yl]ethan-1-one, tech
Catalog # APOSOR32906-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :237.705 g/mol
Storage Temperature:Ambient
CAS Number:672950-24-2
Formula:C₁₁H₈ClNOS
MDL Number:MFCD02187036