Order Entry
Italy
ContactUsLinkComponent
5-(3-Chlorophenyl)-2-oxo-1-(prop-2-en-1-yl)-1,2-dihydropyridine-3-carbonitrile, tech
5-(3-Chlorophenyl)-2-oxo-1-(prop-2-en-1-yl)-1,2-dihydropyridine-3-carbonitrile, tech
Catalog # APOSOR32857-1G
Supplier:  Apollo Scientific
CAS Number:  
5-(3-Chlorophenyl)-2-oxo-1-(prop-2-en-1-yl)-1,2-dihydropyridine-3-carbonitrile, tech
Catalog # APOSOR32857-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :270.714 g/mol
Storage Temperature:Ambient
CAS Number:338954-89-5
Formula:C₁₅H₁₁ClN₂O
MDL Number:MFCD00231597