Order Entry
Italy
ContactUsLinkComponent
6'-Phenyl-2,4-dihydrospiro[1-benzopyran-3,3'-[1,5]dioxane]-4-one, tech
6'-Phenyl-2,4-dihydrospiro[1-benzopyran-3,3'-[1,5]dioxane]-4-one, tech
Catalog # APOSOR32836-1G
Supplier:  Apollo Scientific
CAS Number:  
6'-Phenyl-2,4-dihydrospiro[1-benzopyran-3,3'-[1,5]dioxane]-4-one, tech
Catalog # APOSOR32836-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :296.317 g/mol
Storage Temperature:Ambient
CAS Number:338755-65-0
Formula:C₁₈H₁₆O₄
MDL Number:MFCD00139868