Order Entry
Italy
ContactUsLinkComponent
4-(Piperidin-1-yl)-1lambda6-thiomorpholine-1,1-dione, tech
4-(Piperidin-1-yl)-1lambda6-thiomorpholine-1,1-dione, tech
Catalog # APOSOR32803-1G
Supplier:  Apollo Scientific
CAS Number:  
4-(Piperidin-1-yl)-1lambda6-thiomorpholine-1,1-dione, tech
Catalog # APOSOR32803-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :218.316 g/mol
Storage Temperature:Ambient
CAS Number:866020-54-4
Formula:C₉H₁₈N₂O₂S
MDL Number:MFCD04125551