Order Entry
Italy
ContactUsLinkComponent
4-(Morpholin-4-yl)-1lambda6-thiomorpholine-1,1-dione, tech
4-(Morpholin-4-yl)-1lambda6-thiomorpholine-1,1-dione, tech
Catalog # APOSOR32801-1G
Supplier:  Apollo Scientific
CAS Number:  
4-(Morpholin-4-yl)-1lambda6-thiomorpholine-1,1-dione, tech
Catalog # APOSOR32801-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :220.289 g/mol
Storage Temperature:Ambient
CAS Number:866020-38-4
Formula:C₈H₁₆N₂O₃S
MDL Number:MFCD04125544