Order Entry
Italy
ContactUsLinkComponent
5-Bromo-4,6-dimethyl-2-(morpholin-4-yl)pyridine-3-carbonitrile, tech
5-Bromo-4,6-dimethyl-2-(morpholin-4-yl)pyridine-3-carbonitrile, tech
Catalog # APOSOR32789-1G
Supplier:  Apollo Scientific
CAS Number:  
5-Bromo-4,6-dimethyl-2-(morpholin-4-yl)pyridine-3-carbonitrile, tech
Catalog # APOSOR32789-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :296.163 g/mol
Storage Temperature:Ambient
CAS Number:866019-05-8
Formula:C₁₂H₁₄BrN₃O
MDL Number:MFCD03787140