Order Entry
Italy
ContactUsLinkComponent
2-[(1H-Indol-3-yl)methylidene]-5,5-dimethylcyclohexane-1,3-dione, tech
2-[(1H-Indol-3-yl)methylidene]-5,5-dimethylcyclohexane-1,3-dione, tech
Catalog # APOSOR32771-1G
Supplier:  Apollo Scientific
CAS Number:  
2-[(1H-Indol-3-yl)methylidene]-5,5-dimethylcyclohexane-1,3-dione, tech
Catalog # APOSOR32771-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :267.322 g/mol
Storage Temperature:Ambient
CAS Number:40302-35-0
Formula:C₁₇H₁₇NO₂
MDL Number:MFCD00427871