Order Entry
Italy
ContactUsLinkComponent
5,6-Dimethyl-1-propyl-1H-1,3-benzodiazol-4-amine, tech
5,6-Dimethyl-1-propyl-1H-1,3-benzodiazol-4-amine, tech
Catalog # APOSOR32741-1G
Supplier:  Apollo Scientific
CAS Number:  
5,6-Dimethyl-1-propyl-1H-1,3-benzodiazol-4-amine, tech
Catalog # APOSOR32741-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :203.283 g/mol
Storage Temperature:Ambient
CAS Number:338423-42-0
Formula:C₁₂H₁₇N₃
MDL Number:MFCD00231537