Order Entry
Italy
ContactUsLinkComponent
Ethyl 8-methoxy-2H-chromene-3-carboxylate, tech
Ethyl 8-methoxy-2H-chromene-3-carboxylate, tech
Catalog # APOSOR32726-1G
Supplier:  Apollo Scientific
CAS Number:  
Ethyl 8-methoxy-2H-chromene-3-carboxylate, tech
Catalog # APOSOR32726-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :234.248 g/mol
Storage Temperature:Ambient
CAS Number:112904-72-0
Formula:C₁₃H₁₄O₄
MDL Number:MFCD01316444