Order Entry
Italy
ContactUsLinkComponent
N-Methyl-5H-pyrimido[5,4-b]indol-4-amine, tech
N-Methyl-5H-pyrimido[5,4-b]indol-4-amine, tech
Catalog # APOSOR32720-1G
Supplier:  Apollo Scientific
CAS Number:  
N-Methyl-5H-pyrimido[5,4-b]indol-4-amine, tech
Catalog # APOSOR32720-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :198.224 g/mol
Storage Temperature:Ambient
CAS Number:338419-65-1
Formula:C₁₁H₁₀N₄
MDL Number:MFCD00141078