
7-Bromo-1H-indole-3-carboxaldehyde
Catalog # APOSOR3269-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:224.054 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:115666-21-2
Formula:C₉H₆BrNO
MDL Number:MFCD06657151