Order Entry
Italy
ContactUsLinkComponent
Pyrrolo[1,2-a]quinolin-6-amine, tech
Pyrrolo[1,2-a]quinolin-6-amine, tech
Catalog # APOSOR32687-1G
Supplier:  Apollo Scientific
CAS Number:  
Pyrrolo[1,2-a]quinolin-6-amine, tech
Catalog # APOSOR32687-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :182.221 g/mol
Storage Temperature:Ambient
CAS Number:865658-88-4
Formula:C₁₂H₁₀N₂
MDL Number:MFCD03848484