1-Methyl-2-(morpholin-4-yl)-1H-indole-3-carbaldehyde, tech
Catalog # APOSOR32657-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:244.289 g/mol
Storage Temperature:Ambient
CAS Number:861212-44-4
Formula:C₁₄H₁₆N₂O₂
MDL Number:MFCD04125124