Order Entry
Italy
ContactUsLinkComponent
1-(4-Chlorophenyl)pentane-1,4-dione, tech
1-(4-Chlorophenyl)pentane-1,4-dione, tech
Catalog # APOSOR32652-1G
Supplier:  Apollo Scientific
CAS Number:  
1-(4-Chlorophenyl)pentane-1,4-dione, tech
Catalog # APOSOR32652-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :210.657 g/mol
Storage Temperature:Ambient
CAS Number:53842-12-9
Formula:C₁₁H₁₁ClO₂
MDL Number:MFCD01316205