Order Entry
Italy
Orders LinkContactUsLinkComponent
 

(4-Chlorophenyl)methyl acetate, tech

Supplier:   Apollo Scientific
(4-Chlorophenyl)methyl acetate, tech
Supplier:  Apollo Scientific
Certificates
(4-Chlorophenyl)methyl acetate, tech

3 Options Available Below

Product Details & Documents
Synonyms:  p-Chlorobenzyl acetate, 4-Chlorobenzyl acetate
  • CAS Number:5406-33-7
  • Formula:C₉H₉ClO₂
  • MDL Number:MFCD00027406
  • Molecular Weight:184.62 g/mol
  • Storage Temperature:Ambient
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Bottle
    Log in for availability
    Log in for availability
    1 * 100 mg
    1 * 100 mg - 254,00 €
    254,00 €
    1 * 100 mg
       254,00 €

  • Bottle
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 1 010,00 €
    1 010,00 €
    1 * 1 g
       1 010,00 €

  • Bottle
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 3 030,00 €
    3 030,00 €
    1 * 5 g
       3 030,00 €