Order Entry
Italy
ContactUsLinkComponent
4-Chloro-1-(4-chlorophenyl)-6-oxo-1,6-dihydropyridazine-3-carbonitrile, tech
4-Chloro-1-(4-chlorophenyl)-6-oxo-1,6-dihydropyridazine-3-carbonitrile, tech
Catalog # APOSOR32614-1G
Supplier:  Apollo Scientific
CAS Number:  
4-Chloro-1-(4-chlorophenyl)-6-oxo-1,6-dihydropyridazine-3-carbonitrile, tech
Catalog # APOSOR32614-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :266.083 g/mol
Storage Temperature:Ambient
CAS Number:338405-10-0
Formula:C₁₁H₅Cl₂N₃O
MDL Number:MFCD00141174