Order Entry
Italy
ContactUsLinkComponent
(2E)-3-(Dimethylamino)-1-[4-(morpholin-4-yl)phenyl]prop-2-en-1-one, tech
(2E)-3-(Dimethylamino)-1-[4-(morpholin-4-yl)phenyl]prop-2-en-1-one, tech
Catalog # APOSOR32587-1G
Supplier:  Apollo Scientific
CAS Number:  
(2E)-3-(Dimethylamino)-1-[4-(morpholin-4-yl)phenyl]prop-2-en-1-one, tech
Catalog # APOSOR32587-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :260.332 g/mol
Storage Temperature:Ambient
CAS Number:1345865-34-0
Formula:C₁₅H₂₀N₂O₂
MDL Number:MFCD03617857