Order Entry
Italy
ContactUsLinkComponent
N-(4-Chlorophenyl)-1-oxa-4-azaspiro[4.5]decane-4-carboxamide, tech
N-(4-Chlorophenyl)-1-oxa-4-azaspiro[4.5]decane-4-carboxamide, tech
Catalog # APOSOR32577-1G
Supplier:  Apollo Scientific
CAS Number:  
N-(4-Chlorophenyl)-1-oxa-4-azaspiro[4.5]decane-4-carboxamide, tech
Catalog # APOSOR32577-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :294.777 g/mol
Storage Temperature:Ambient
CAS Number:861209-90-7
Formula:C₁₅H₁₉ClN₂O₂
MDL Number:MFCD03618017