1H,2H-Naphtho[2,1-b]furan-1-one, tech
Catalog # APOSOR32551-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:184.191 g/mol
Storage Temperature:Ambient
CAS Number:19997-42-3
Formula:C₁₂H₈O₂
MDL Number:MFCD00975011