Ethyl 2-(2-oxo-1,2-dihydroquinoxalin-1-yl)acetate, tech
Catalog # APOSOR32543-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:232.235 g/mol
Storage Temperature:Ambient
CAS Number:154640-54-7
Formula:C₁₂H₁₂N₂O₃
MDL Number:MFCD05155084