Order Entry
Italy
ContactUsLinkComponent
 
2-(Morpholin-4-yl)-1,1-diphenylethan-1-ol, tech
Catalog # APOSOR32540-1G
Supplier:  Apollo Scientific
CAS Number:  
2-(Morpholin-4-yl)-1,1-diphenylethan-1-ol, tech
Catalog # APOSOR32540-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :283.365 g/mol
Storage Temperature:Ambient
CAS Number:13150-37-3
Formula:C₁₈H₂₁NO₂
MDL Number:MFCD00231484