Order Entry
Italy
ContactUsLinkComponent
3-Hydroxy-2-methyl-3-(2-phenylethynyl)-2,3-dihydro-1H-isoindol-1-one, tech
3-Hydroxy-2-methyl-3-(2-phenylethynyl)-2,3-dihydro-1H-isoindol-1-one, tech
Catalog # APOSOR32535-1G
Supplier:  Apollo Scientific
CAS Number:  
3-Hydroxy-2-methyl-3-(2-phenylethynyl)-2,3-dihydro-1H-isoindol-1-one, tech
Catalog # APOSOR32535-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :263.291 g/mol
Storage Temperature:Ambient
CAS Number:143647-54-5
Formula:C₁₇H₁₃NO₂
MDL Number:MFCD00215773