N-[4-(2-Phenylethynyl)phenyl]methanesulphonamide, tech
Catalog # APOSOR32482-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:271.334 g/mol
Storage Temperature:Ambient
CAS Number:439095-76-8
Formula:C₁₅H₁₃NO₂S
MDL Number:MFCD03012868