Order Entry
Italy
ContactUsLinkComponent
2-Chloro-3-(pyrrolidin-1-yl)-1,4-dihydronaphthalene-1,4-dione, tech
2-Chloro-3-(pyrrolidin-1-yl)-1,4-dihydronaphthalene-1,4-dione, tech
Catalog # APOSOR32470-1G
Supplier:  Apollo Scientific
CAS Number:  
2-Chloro-3-(pyrrolidin-1-yl)-1,4-dihydronaphthalene-1,4-dione, tech
Catalog # APOSOR32470-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :261.704 g/mol
Storage Temperature:Ambient
CAS Number:59641-25-7
Formula:C₁₄H₁₂ClNO₂
MDL Number:MFCD00010368