Order Entry
Italy
ContactUsLinkComponent
1,5-Dimethyl-4-phenylimidazolidin-2-one 95%
1,5-Dimethyl-4-phenylimidazolidin-2-one 95%
Catalog # APOSOR32468-1G
Supplier:  Apollo Scientific
CAS Number:  
1,5-Dimethyl-4-phenylimidazolidin-2-one 95%
Catalog # APOSOR32468-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :190.242 g/mol
Storage Temperature:Ambient
CAS Number:103774-40-9
Formula:C₁₁H₁₄N₂O
MDL Number:MFCD01456559