Order Entry
Italy
ContactUsLinkComponent
6-(3-Chlorophenyl)-2-methylpyrazolo[1,5-a]pyrimidin-7-amine, tech
6-(3-Chlorophenyl)-2-methylpyrazolo[1,5-a]pyrimidin-7-amine, tech
Catalog # APOSOR32467-1G
Supplier:  Apollo Scientific
CAS Number:  
6-(3-Chlorophenyl)-2-methylpyrazolo[1,5-a]pyrimidin-7-amine, tech
Catalog # APOSOR32467-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :258.706 g/mol
Storage Temperature:Ambient
CAS Number:85841-03-8
Formula:C₁₃H₁₁ClN₄
MDL Number:MFCD03102690