Order Entry
Italy
ContactUsLinkComponent
1-[(2-Methyl-1H-indol-4-yl)oxy]-3-(morpholin-4-yl)propan-2-ol, tech
1-[(2-Methyl-1H-indol-4-yl)oxy]-3-(morpholin-4-yl)propan-2-ol, tech
Catalog # APOSOR32431-1G
Supplier:  Apollo Scientific
CAS Number:  
1-[(2-Methyl-1H-indol-4-yl)oxy]-3-(morpholin-4-yl)propan-2-ol, tech
Catalog # APOSOR32431-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :290.357 g/mol
Storage Temperature:Ambient
CAS Number:338392-10-2
Formula:C₁₆H₂₂N₂O₃
MDL Number:MFCD01315611