Ethyl 2-[(3-methylphenyl)formamido]acetate, tech
Catalog # APOSOR32424-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:221.252 g/mol
Storage Temperature:Ambient
CAS Number:330466-40-5
Formula:C₁₂H₁₅NO₃
MDL Number:MFCD00793749