Order Entry
Italy
ContactUsLinkComponent
Methyl (4Z)-4-[(dimethylamino)methylidene]-2-methyl-5-oxo-4,5-dihydro-1H-pyrrole-3-carboxylate, tech
Methyl (4Z)-4-[(dimethylamino)methylidene]-2-methyl-5-oxo-4,5-dihydro-1H-pyrrole-3-carboxylate, tech
Catalog # APOSOR32413-1G
Supplier:  Apollo Scientific
CAS Number:  
Methyl (4Z)-4-[(dimethylamino)methylidene]-2-methyl-5-oxo-4,5-dihydro-1H-pyrrole-3-carboxylate, tech
Catalog # APOSOR32413-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :210.23 g/mol
Storage Temperature:Ambient
CAS Number:477865-31-9
Formula:C₁₀H₁₄N₂O₃
MDL Number:MFCD00244306