Inserisci ordine
Italy
ContactUsLinkComponent
(3E)-2-(Morpholin-4-yl)-4-phenylbut-3-enenitrile, tech
(3E)-2-(Morpholin-4-yl)-4-phenylbut-3-enenitrile, tech
Catalog # APOSOR32405-1G
Supplier:  Apollo Scientific
CAS Number:  
(3E)-2-(Morpholin-4-yl)-4-phenylbut-3-enenitrile, tech
Catalog # APOSOR32405-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Tipo di conf.:
    Bottle
  • Conf:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :228.29 g/mol
Storage Temperature:Ambiente
CAS Number:101132-14-3
Formula:C₁₄H₁₆N₂O
MDL Number:MFCD00173087