3-(4-Methoxyphenoxy)butan-2-one, tech
Catalog # APOSOR32400-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:194.227 g/mol
Storage Temperature:Ambient
CAS Number:24264-51-5
Formula:C₁₁H₁₄O₃
MDL Number:MFCD00231719