5,6-Dibromocyclohex-2-ene-1,4-dione, tech
Catalog # APOSOR32323-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:267.903 g/mol
Storage Temperature:Ambient
CAS Number:5273-61-0
Formula:C₆H₄Br₂O₂
MDL Number:MFCD01568777