Order Entry
Italy
ContactUsLinkComponent
Ethyl 1-(4-chlorophenyl)-5-methyl-1H-1,2,4-triazole-3-carboxylate, tech
Ethyl 1-(4-chlorophenyl)-5-methyl-1H-1,2,4-triazole-3-carboxylate, tech
Catalog # APOSOR32299-1G
Supplier:  Apollo Scientific
CAS Number:  
Ethyl 1-(4-chlorophenyl)-5-methyl-1H-1,2,4-triazole-3-carboxylate, tech
Catalog # APOSOR32299-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :265.696 g/mol
Storage Temperature:Ambient
CAS Number:91396-27-9
Formula:C₁₂H₁₂ClN₃O₂
MDL Number:MFCD00173065