Order Entry
Italy
ContactUsLinkComponent
7-Chloro-n-p-tolylthiazolo[5,4-d]pyrimidin-2-amine
7-Chloro-n-p-tolylthiazolo[5,4-d]pyrimidin-2-amine
Catalog # APOSOR322974-250MG
Supplier:  Apollo Scientific
CAS Number:  
7-Chloro-n-p-tolylthiazolo[5,4-d]pyrimidin-2-amine
Catalog # APOSOR322974-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :276.745 g/mol
Storage Temperature:Ambient
CAS Number:871266-80-7
Formula:C₁₂H₉ClN₄S
MDL Number:MFCD08437191