3-Acetyl-4-hydroxy-1-phenyl-1,2-dihydroquinolin-2-one, tech
Catalog # APOSOR32275-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:279.29 g/mol
Storage Temperature:Ambient
CAS Number:54289-77-9
Formula:C₁₇H₁₃NO₃
MDL Number:MFCD00179942