Order Entry
Italy
ContactUsLinkComponent
 
3-Bromo-4-(morpholin-4-yl)aniline 95%
Catalog # APOSOR32233-1G
Supplier:  Apollo Scientific
CAS Number:  
3-Bromo-4-(morpholin-4-yl)aniline 95%
Catalog # APOSOR32233-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :257.127 g/mol
Storage Temperature:Ambient
CAS Number:347324-26-9
Formula:C₁₀H₁₃BrN₂O
MDL Number:MFCD02081966