Methyl 1-(4-chlorobenzoyl)piperidine-4-carboxylate, tech
Catalog # APOSOR32226-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:281.735 g/mol
Storage Temperature:Ambient
CAS Number:349614-05-7
Formula:C₁₄H₁₆ClNO₃
MDL Number:MFCD02082485