Order Entry
Italy
ContactUsLinkComponent
5-Chloro-1,3-dimethyl-1H-pyrazole-4-carbonitrile 95%
5-Chloro-1,3-dimethyl-1H-pyrazole-4-carbonitrile 95%
Catalog # APOSOR32223-1G
Supplier:  Apollo Scientific
CAS Number:  
5-Chloro-1,3-dimethyl-1H-pyrazole-4-carbonitrile 95%
Catalog # APOSOR32223-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :155.585 g/mol
Storage Temperature:Ambient
CAS Number:96286-02-1
Formula:C₆H₆ClN₃
MDL Number:MFCD01569082