Order Entry
Italy
ContactUsLinkComponent
4,5-Dichloro-2-ethyl-2,3-dihydropyridazin-3-one 95%
4,5-Dichloro-2-ethyl-2,3-dihydropyridazin-3-one 95%
Catalog # APOSOR32207-1G
Supplier:  Apollo Scientific
CAS Number:  
4,5-Dichloro-2-ethyl-2,3-dihydropyridazin-3-one 95%
Catalog # APOSOR32207-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :193.031 g/mol
Storage Temperature:Ambient
CAS Number:33098-10-1
Formula:C₆H₆Cl₂N₂O
MDL Number:MFCD00793799