3-Acetyl-4-hydroxy-1-azatricyclo[7.3.1.05,13]trideca-3,5(13),6,8-tetraen-2-one, tech
Catalog # APOSOR32155-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:243.258 g/mol
Storage Temperature:Ambient
CAS Number:141945-41-7
Formula:C₁₄H₁₃NO₃
MDL Number:MFCD02178524