Order Entry
Italy
ContactUsLinkComponent
10-Acetyl-9-hydroxy-1-azatricyclo[6.3.1.04,12]dodeca-4,6,8(12),9-tetraen-11-one, tech
10-Acetyl-9-hydroxy-1-azatricyclo[6.3.1.04,12]dodeca-4,6,8(12),9-tetraen-11-one, tech
Catalog # APOSOR32150-1G
Supplier:  Apollo Scientific
CAS Number:  
10-Acetyl-9-hydroxy-1-azatricyclo[6.3.1.04,12]dodeca-4,6,8(12),9-tetraen-11-one, tech
Catalog # APOSOR32150-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :229.231 g/mol
Storage Temperature:Ambient
CAS Number:128099-76-3
Formula:C₁₃H₁₁NO₃
MDL Number:MFCD01566965