Ethyl 3-chloroimidazo[1,2-a]pyrimidine-2-carboxylate, tech
Codice catalogo APOSOR32138-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:225.632 g/mol
Storage Temperature:Ambient
CAS Number:860612-52-8
Formula:C₉H₈ClN₃O₂
MDL Number:MFCD04124241