Order Entry
Italy
ContactUsLinkComponent
tert-Butyl 4,7-diazaspiro[2.5]octane-4-carboxylate 97%
Irritant
tert-Butyl 4,7-diazaspiro[2.5]octane-4-carboxylate 97%
Catalog # APOSOR321367-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
tert-Butyl 4,7-diazaspiro[2.5]octane-4-carboxylate 97%
Catalog # APOSOR321367-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :212.289 g/mol
 :GHS07
Storage Temperature:Refrigerator
CAS Number:674792-08-6
Formula:C₁₁H₂₀N₂O₂
MDL Number:MFCD08685932