
Methyl 2-(piperidin-4-yl)benzoate hydrochloride 97%
Catalog # APOSOR321223-100MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
:255.741 g/mol
:GHS07
Storage Temperature:Refrigerator
CAS Number:176526-08-2
Formula:C₁₃H₁₇NO₂·ClH
MDL Number:MFCD02179143