Ethyl 2-(naphthalen-1-yloxy)acetate, tech
Catalog # APOSOR32121-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:230.259 g/mol
Storage Temperature:Ambient
CAS Number:41643-81-6
Formula:C₁₄H₁₄O₃
MDL Number:MFCD01162455