Order Entry
Italy
ContactUsLinkComponent
[1-(Methylamino)-1H,2H,3H-naphtho[2,1-b]pyran-2-yl]methanol hydrochloride, tech
[1-(Methylamino)-1H,2H,3H-naphtho[2,1-b]pyran-2-yl]methanol hydrochloride, tech
Catalog # APOSOR32065-1G
Supplier:  Apollo Scientific
CAS Number:  
[1-(Methylamino)-1H,2H,3H-naphtho[2,1-b]pyran-2-yl]methanol hydrochloride, tech
Catalog # APOSOR32065-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :279.762 g/mol
Storage Temperature:Ambient
CAS Number:477708-75-1
Formula:C₁₅H₁₇NO₂·ClH
MDL Number:MFCD00663770